Assay Details
| Gene Symbol: eif3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016190.2 | NP_001016190.1 | 3 - 4 | 313 | 63 | |
| GenBank mRNA | CR761801.2 | - | 3 - 4 | 313 | 63 | |
More Information
Set Membership: |
|

eif3f
Genomic Map