Assay Details
| Gene Symbol: cop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016199.2 | NP_001016199.1 | 16 - 17 | 1816 | 84 | |
| XM_012961071.2 | XP_012816525.1 | 16 - 17 | 2332 | 84 | ||
| XM_018093039.1 | XP_017948528.1 | 13 - 14 | 1312 | 84 | ||
| GenBank mRNA | BC170678.1 | - | 16 - 17 | 1748 | 84 | |
| CR926346.2 | - | 16 - 17 | 1816 | 84 | ||
More Information
Set Membership: |
|

cop1
Genomic Map