Assay Details
| Gene Symbol: pigu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016206.2 | NP_001016206.1 | 2 - 3 | 252 | 84 | |
| XM_012971762.2 | XP_012827216.1 | 2 - 3 | 364 | 84 | ||
| GenBank mRNA | BC170796.1 | - | 2 - 3 | 222 | 84 | |
| BC170798.1 | - | 2 - 3 | 222 | 84 | ||
| CR926339.2 | - | 2 - 3 | 252 | 84 | ||
More Information
Set Membership: |
|

pigu
Genomic Map