Assay Details
| Gene Symbol: pdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016212.2 | NP_001016212.1 | 9 - 10 | 944 | 63 | |
| GenBank mRNA | BC091061.1 | - | 9 - 10 | 944 | 63 | |
| CR926329.2 | - | 9 - 10 | 936 | 63 | ||
More Information
Set Membership: |
|

pdhb
Genomic Map