Assay Details
| Gene Symbol: dkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016283.2 | NP_001016283.1 | 2 - 3 | 505 | 67 | |
| GenBank mRNA | BC157321.1 | - | 2 - 3 | 507 | 67 | |
| CR761605.2 | - | 2 - 3 | 505 | 67 | ||
More Information
Set Membership: |
|

dkk1
Genomic Map