Assay Details
Gene Symbol: dcun1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016315.2 | NP_001016315.1 | 6 - 7 | 1187 | 113 | |
GenBank mRNA | BC091083.1 | - | 5 - 6 | 690 | 113 | |
BC167342.1 | - | 7 - 8 | 936 | 113 | ||
BC170696.1 | - | 7 - 8 | 1024 | 113 | ||
BC171114.1 | - | 7 - 8 | 1024 | 113 | ||
CR761471.2 | - | 6 - 7 | 1187 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
