Assay Details
| Gene Symbol: dusp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016317.2 | NP_001016317.1 | 1 - 2 | 254 | 61 | |
| GenBank mRNA | BC158155.1 | - | 1 - 2 | 292 | 61 | |
| CR761465.2 | - | 1 - 2 | 254 | 61 | ||
More Information
Set Membership: |
|

dusp19
Genomic Map