Assay Details
Gene Symbol: rsrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016318.3 | NP_001016318.1 | 6 - 7 | 634 | 67 | |
XM_012962980.2 | XP_012818434.1 | 7 - 8 | 742 | 67 | ||
GenBank mRNA | BC157777.1 | - | 7 - 8 | 689 | 67 | |
CR761462.2 | - | 6 - 7 | 637 | 67 | ||
CT027861.1 | - | 7 - 8 | 687 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
