Assay Details
| Gene Symbol: snx33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016368.2 | NP_001016368.1 | 2 - 3 | 1175 | 97 | |
| XM_012966575.2 | XP_012822029.1 | 2 - 3 | 961 | 97 | ||
| XM_018092084.1 | XP_017947573.1 | 2 - 3 | 1091 | 97 | ||
| GenBank mRNA | BC127346.1 | - | 2 - 3 | 931 | 97 | |
| CR761284.2 | - | 2 - 3 | 1175 | 97 | ||
More Information
Set Membership: |
|

snx33
Genomic Map