Assay Details
| Gene Symbol: lsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016402.3 | NP_001016402.1 | 3 - 4 | 235 | 110 | |
| GenBank mRNA | CN108372.1 | - | 3 - 4 | 237 | 110 | |
| CR761142.1 | - | 3 - 4 | 114 | 110 | ||
| DN095519.1 | - | 3 - 4 | 158 | 110 | ||
| DR900519.1 | - | 3 - 4 | 207 | 110 | ||
More Information
Set Membership: |
|

lsm2
Genomic Map