Assay Details
| Gene Symbol: ucma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016485.2 | NP_001016485.1 | 3 - 4 | 223 | 74 | |
| GenBank mRNA | BC135882.1 | - | 3 - 4 | 237 | 74 | |
| CR760848.2 | - | 3 - 4 | 223 | 74 | ||
More Information
Set Membership: |
|

ucma
Genomic Map