Assay Details
Gene Symbol: vdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016492.2 | NP_001016492.1 | 5 - 6 | 342 | 79 | |
GenBank mRNA | BC127348.1 | - | 4 - 5 | 336 | 79 | |
BC135545.1 | - | 4 - 5 | 317 | 79 | ||
CR760826.2 | - | 5 - 6 | 342 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
