Assay Details
| Gene Symbol: snap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016501.2 | NP_001016501.1 | 4 - 5 | 277 | 64 | |
| GenBank mRNA | BC077066.1 | - | 4 - 5 | 311 | 64 | |
| BC135565.1 | - | 4 - 5 | 294 | 64 | ||
| CR760797.2 | - | 4 - 5 | 277 | 64 | ||
More Information
Set Membership: |
|

snap25
Genomic Map