Assay Details
| Gene Symbol: pou3f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016504.2 | NP_001016504.1 | 1 | 1051 | 98 | |
| GenBank mRNA | BC098088.1 | - | 1 | 1051 | 98 | |
| CR760792.2 | - | 1 | 1027 | 98 | ||
More Information
Set Membership: |
|

pou3f1
Genomic Map