Assay Details
| Gene Symbol: suv39h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016508.1 | NP_001016508.1 | 3 - 4 | 852 | 108 | |
| XM_012959025.2 | XP_012814479.1 | 3 - 4 | 1776 | 108 | ||
| GenBank mRNA | BC158299.1 | - | 3 - 4 | 971 | 108 | |
| CR926249.1 | - | 3 - 4 | 852 | 108 | ||
More Information
Set Membership: |
|

suv39h2
Genomic Map