Assay Details
Gene Symbol: atp5f1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016516.2 | NP_001016516.1 | 1 - 2 | 146 | 63 | |
GenBank mRNA | BC135474.1 | - | 1 - 2 | 139 | 63 | |
CR926220.2 | - | 1 - 2 | 146 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
