Assay Details
Gene Symbol: noct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016531.1 | NP_001016531.1 | 1 - 2 | 376 | 82 | |
GenBank mRNA | BC170786.1 | - | 1 - 2 | 371 | 82 | |
CR926178.1 | - | 1 - 2 | 376 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
