Assay Details
| Gene Symbol: noct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016531.1 | NP_001016531.1 | 2 - 3 | 649 | 92 | |
| GenBank mRNA | BC170786.1 | - | 2 - 3 | 644 | 92 | |
| CR926178.1 | - | 2 - 3 | 649 | 92 | ||
More Information
Set Membership: |
|

noct
Genomic Map