Assay Details
| Gene Symbol: rpl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016534.1 | NP_001016534.1 | 3 - 4 | 313 | 75 | |
| XM_018094654.1 | XP_017950143.1 | 4 - 5 | 713 | 75 | ||
| GenBank mRNA | CR926169.1 | - | 3 - 4 | 313 | 75 | |
More Information
Set Membership: |
|

rpl14
Genomic Map