Assay Details
| Gene Symbol: rps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016566.2 | NP_001016566.1 | 4 - 5 | 517 | 65 | |
| GenBank mRNA | BC118703.1 | - | 5 - 6 | 525 | 65 | |
| CR760677.2 | - | 4 - 5 | 517 | 65 | ||
| CT025346.2 | - | 4 - 5 | 520 | 65 | ||
More Information
Set Membership: |
|

rps8
Genomic Map