Assay Details
| Gene Symbol: pitpnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016639.2 | NP_001016639.1 | 8 - 9 | 775 | 65 | |
| XM_012966615.2 | XP_012822069.1 | 8 - 9 | 911 | 65 | ||
| XM_018095393.1 | XP_017950882.1 | 8 - 9 | 914 | 65 | ||
| GenBank mRNA | BC118687.1 | - | 8 - 9 | 715 | 65 | |
| BC171116.1 | - | 8 - 9 | 702 | 65 | ||
| CR760415.2 | - | 8 - 9 | 775 | 65 | ||
More Information
Set Membership: |
|

pitpnc1
Genomic Map