Assay Details
| Gene Symbol: rplp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016645.2 | NP_001016645.1 | 2 - 3 | 147 | 77 | |
| GenBank mRNA | BC135754.1 | - | 2 - 3 | 210 | 77 | |
| CR760396.2 | - | 2 - 3 | 147 | 77 | ||
More Information
Set Membership: |
|

rplp2
Genomic Map