Assay Details
Gene Symbol: txnl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016646.2 | NP_001016646.1 | 2 - 3 | 274 | 78 | |
GenBank mRNA | BC157181.1 | - | 3 - 4 | 346 | 78 | |
CR760391.2 | - | 2 - 3 | 274 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
