Assay Details
| Gene Symbol: gsta4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016676.2 | NP_001016676.1 | 6 - 7 | 596 | 101 | |
| XM_018093890.1 | XP_017949379.1 | 7 - 8 | 757 | 101 | ||
| GenBank mRNA | BC170614.1 | - | 6 - 7 | 594 | 101 | |
| BC170620.1 | - | 6 - 7 | 594 | 101 | ||
| CR760229.2 | - | 6 - 7 | 596 | 101 | ||
More Information
Set Membership: |
|

gsta4
Genomic Map