Assay Details
| Gene Symbol: rpl18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016690.2 | NP_001016690.1 | 2 - 3 | 204 | 112 | |
| GenBank mRNA | BC161335.1 | - | 1 - 2 | 167 | 112 | |
| BC167567.1 | - | 2 - 3 | 214 | 112 | ||
| BC167657.1 | - | 1 - 2 | 159 | 112 | ||
| CR760185.2 | - | 2 - 3 | 204 | 112 | ||
More Information
Set Membership: |
|

rpl18a
Genomic Map