Assay Details
| Gene Symbol: rpl18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016690.2 | NP_001016690.1 | 3 - 4 | 334 | 103 | |
| GenBank mRNA | BC135797.1 | - | 3 - 4 | 342 | 103 | |
| BC161335.1 | - | 2 - 3 | 297 | 103 | ||
| BC167567.1 | - | 3 - 4 | 344 | 103 | ||
| BC167657.1 | - | 2 - 3 | 289 | 103 | ||
| CR760185.2 | - | 3 - 4 | 334 | 103 | ||
More Information
Set Membership: |
|

rpl18a
Genomic Map