Assay Details
Gene Symbol: znf706.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016744.1 | NP_001016744.1 | 2 - 3 | 165 | 80 | |
XM_012956994.2 | XP_012812448.1 | 3 - 4 | 384 | 80 | ||
XM_012956996.2 | XP_012812450.1 | 2 - 3 | 222 | 80 | ||
XM_012956997.2 | XP_012812451.1 | 2 - 3 | 226 | 80 | ||
GenBank mRNA | BC157779.1 | - | 2 - 3 | 243 | 80 | |
CR759984.1 | - | 2 - 3 | 165 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
