Assay Details
Gene Symbol: pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016953.2 | NP_001016953.1 | 4 - 5 | 784 | 83 | |
GenBank mRNA | CR761607.2 | - | 4 - 5 | 784 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
