Assay Details
Gene Symbol: rab4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016987.2 | NP_001016987.1 | 2 - 3 | 120 | 99 | |
XM_018096270.1 | XP_017951759.1 | 2 - 3 | 274 | 99 | ||
XM_018096271.1 | XP_017951760.1 | 2 - 3 | 279 | 99 | ||
GenBank mRNA | BC168000.1 | - | 2 - 3 | 247 | 99 | |
CR848260.2 | - | 2 - 3 | 120 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
