Assay Details
| Gene Symbol: rps5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016992.3 | NP_001016992.1 | 3 - 4 | 444 | 62 | |
| XM_012968458.1 | XP_012823912.1 | 4 - 5 | 640 | 62 | ||
| GenBank mRNA | BC082738.1 | - | 4 - 5 | 494 | 62 | |
| BC135112.1 | - | 4 - 5 | 478 | 62 | ||
| BC153694.1 | - | 3 - 4 | 454 | 62 | ||
| BC161332.1 | - | 3 - 4 | 445 | 62 | ||
| CR848217.2 | - | 3 - 4 | 446 | 62 | ||
More Information
Set Membership: |
|

rps5
Genomic Map