Assay Details
| Gene Symbol: ss18l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017009.2 | NP_001017009.1 | 1 - 2 | 95 | 61 | |
| GenBank mRNA | CR762275.2 | - | 1 - 2 | 95 | 61 | |
More Information
Set Membership: |
|

ss18l2
Genomic Map