Assay Details
| Gene Symbol: slc25a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017018.3 | NP_001017018.1 | 4 - 5 | 432 | 61 | |
| GenBank mRNA | CR762144.2 | - | 4 - 5 | 381 | 61 | |
| DR891561.1 | - | 4 - 5 | 432 | 61 | ||
More Information
Set Membership: |
|

slc25a10
Genomic Map