Assay Details
Gene Symbol: pskh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017030.2 | NP_001017030.1 | 1 - 2 | 1034 | 93 | |
XM_012961168.2 | XP_012816622.1 | 2 - 3 | 1095 | 93 | ||
GenBank mRNA | BC157265.1 | - | 1 - 2 | 1011 | 93 | |
CR762032.1 | - | 1 - 2 | 1034 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
