Assay Details
| Gene Symbol: ppib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017063.2 | NP_001017063.1 | 2 - 3 | 231 | 65 | |
| GenBank mRNA | CR761630.2 | - | 2 - 3 | 231 | 65 | |
More Information
Set Membership: |
|

ppib
Genomic Map