Assay Details
Gene Symbol: ebna1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017151.2 | NP_001017151.1 | 5 - 6 | 480 | 68 | |
GenBank mRNA | BC159010.1 | - | 5 - 6 | 476 | 68 | |
CR760915.2 | - | 5 - 6 | 480 | 68 |
Gene Symbol: ebna1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004913957.3 | XP_004914014.1 | 5 - 6 | 548 | 68 | |
GenBank mRNA | CU075329.1 | - | 4 - 5 | 449 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
