Assay Details
| Gene Symbol: ebna1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017151.2 | NP_001017151.1 | 6 - 7 | 572 | 73 | |
| GenBank mRNA | BC159010.1 | - | 6 - 7 | 568 | 73 | |
| CR760915.2 | - | 6 - 7 | 572 | 73 | ||
| Gene Symbol: ebna1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004913957.3 | XP_004914014.1 | 6 - 7 | 640 | 73 | |
| GenBank mRNA | CU075329.1 | - | 5 - 6 | 541 | 73 | |
More Information
Set Membership: |
|

ebna1bp2
Genomic Map