Assay Details
| Gene Symbol: hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017223.2 | NP_001017223.1 | 2 - 3 | 677 | 61 | |
| GenBank mRNA | BC136201.1 | - | 2 - 3 | 744 | 61 | |
| CR760318.2 | - | 2 - 3 | 677 | 61 | ||
More Information
Set Membership: |
|

hspa13
Genomic Map