Assay Details
| Gene Symbol: ndufb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017250.2 | NP_001017250.1 | 2 - 3 | 296 | 101 | |
| GenBank mRNA | BC135549.1 | - | 2 - 3 | 301 | 101 | |
| CR760102.2 | - | 2 - 3 | 296 | 101 | ||
More Information
Set Membership: |
|

ndufb10
Genomic Map