Assay Details
Gene Symbol: lhfpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018091281.1 | XP_017946770.1 | 2 - 3 | 690 | 78 | |
GenBank mRNA | CR855809.2 | - | 4 - 5 | 711 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
