Assay Details
| Gene Symbol: b3gat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030349.1 | NP_001025520.1 | 3 - 4 | 522 | 69 | |
| XM_012954327.2 | XP_012809781.1 | 5 - 6 | 906 | 69 | ||
| XM_012954328.2 | XP_012809782.1 | 4 - 5 | 724 | 69 | ||
| XM_012954329.2 | XP_012809783.1 | 5 - 6 | 894 | 69 | ||
| XM_012954330.2 | XP_012809784.1 | 4 - 5 | 712 | 69 | ||
| XM_012954331.2 | XP_012809785.1 | 4 - 5 | 704 | 69 | ||
| GenBank mRNA | AJ890837.1 | - | 3 - 4 | 522 | 69 | |
| BC121200.1 | - | 5 - 6 | 809 | 69 | ||
More Information
Set Membership: |
|

b3gat3
Genomic Map