Assay Details
| Gene Symbol: gmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030358.1 | NP_001025529.1 | 7 - 8 | 732 | 67 | |
| XM_012963677.1 | XP_012819131.1 | 7 - 8 | 761 | 67 | ||
| XM_012963686.2 | XP_012819140.1 | 7 - 8 | 781 | 67 | ||
| GenBank mRNA | BC090559.1 | - | 7 - 8 | 732 | 67 | |
More Information
Set Membership: |
|

gmpr2
Genomic Map