Assay Details
Gene Symbol: sept12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030371.1 | NP_001025542.1 | 978 | 70 | ||
GenBank mRNA | BC090575.1 | - | 978 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
