Assay Details
Gene Symbol: hmgxb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030384.1 | NP_001025555.1 | 9 - 10 | 1687 | 70 | |
XM_012961211.2 | XP_012816665.1 | 10 - 11 | 2072 | 70 | ||
XM_012961213.2 | XP_012816667.1 | 9 - 10 | 1694 | 70 | ||
GenBank mRNA | BC090595.1 | - | 9 - 10 | 1687 | 70 | |
CR760909.2 | - | 9 - 10 | 1683 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
