Assay Details
Gene Symbol: pkia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030409.1 | NP_001025580.1 | 3 - 4 | 453 | 102 | |
GenBank mRNA | BC091024.1 | - | 3 - 4 | 453 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
