Assay Details
| Gene Symbol: adh1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030410.1 | NP_001025581.1 | 7 - 8 | 1017 | 78 | |
| GenBank mRNA | BC091025.1 | - | 7 - 8 | 1017 | 78 | |
More Information
Set Membership: |
|

adh1c
Genomic Map