Assay Details
| Gene Symbol: aoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030413.1 | NP_001025584.1 | 2 - 3 | 1949 | 74 | |
| GenBank mRNA | BC091028.1 | - | 2 - 3 | 1949 | 74 | |
More Information
Set Membership: |
|

aoc3
Genomic Map