Assay Details
Gene Symbol: sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030422.1 | NP_001025593.1 | 2 - 3 | 244 | 78 | |
XM_012963073.2 | XP_012818527.1 | 2 - 3 | 250 | 78 | ||
GenBank mRNA | BC091042.1 | - | 2 - 3 | 244 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
