Assay Details
| Gene Symbol: fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030424.1 | NP_001025595.1 | 4 - 5 | 569 | 70 | |
| XM_012961218.2 | XP_012816672.1 | 4 - 5 | 720 | 70 | ||
| GenBank mRNA | BC091044.1 | - | 4 - 5 | 569 | 70 | |
More Information
Set Membership: |
|

fmo2
Genomic Map