Assay Details
Gene Symbol: ggh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030427.1 | NP_001025598.1 | 1 - 2 | 242 | 79 | |
XM_012964805.2 | XP_012820259.1 | 1 - 2 | 326 | 79 | ||
GenBank mRNA | BC091047.1 | - | 1 - 2 | 242 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
