Assay Details
| Gene Symbol: atp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030431.1 | NP_001025602.1 | 2 - 3 | 241 | 94 | |
| XM_012959130.2 | XP_012814584.1 | 2 - 3 | 481 | 94 | ||
| GenBank mRNA | BC091052.1 | - | 2 - 3 | 241 | 94 | |
| CR761179.2 | - | 2 - 3 | 216 | 94 | ||
More Information
Set Membership: |
|

atp23
Genomic Map